Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033314.3 | NP_001028486.1 | 8 - 9 | 1283 | 140 | |
GenBank mRNA | AK082980.1 | - | 8 - 9 | 141 | ||
AK146297.1 | - | 8 - 9 | 1284 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |