Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011661.4 | NP_035791.1 | 3 - 4 | 1242 | 73 | |
GenBank mRNA | AK014619.1 | - | 3 - 4 | 1282 | 73 | |
AK148172.1 | - | 3 - 4 | 1265 | 73 | ||
AK148341.1 | - | 3 - 4 | 1192 | 73 | ||
AK148432.1 | - | 3 - 4 | 1221 | 73 | ||
AY526904.1 | - | 3 - 4 | 1283 | 73 | ||
BC079678.1 | - | 3 - 4 | 1192 | 30539846 | 73 | |
D00440.1 | - | 3 - 4 | 1242 | 73 | ||
M20234.1 | - | 3 - 4 | 1343 | 73 | ||
M24560.1 | - | 3 - 4 | 1242 | 73 | ||
M26729.1 | - | 3 - 4 | 1181 | 73 | ||
X12782.1 | - | 3 - 4 | 1283 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |