Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fnbp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114665.2 | NP_001108137.1 | 9 - 10 | 1155 | 95 | |
GenBank mRNA | BM964102.1 | - | 2 - 3 | 233 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |