Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200005164.mm.1 |
NM_001252534.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005164.mm.1 |
NM_001252534.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005164.mm.1 |
NM_001252536.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005164.mm.1 |
NM_001252536.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005164.mm.1 |
NM_001252537.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005164.mm.1 |
NM_001252537.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005164.mm.1 |
NM_001252538.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005164.mm.1 |
NM_001252538.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005164.mm.1 |
NM_028412.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005164.mm.1 |
NM_028412.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005164.mm.1 |
TC0200000577.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200005169.mm.1 |
NM_001252534.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005169.mm.1 |
NM_001252534.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005169.mm.1 |
NM_001252536.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005169.mm.1 |
NM_001252536.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005169.mm.1 |
NM_001252537.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005169.mm.1 |
NM_001252537.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005169.mm.1 |
NM_001252538.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005169.mm.1 |
NM_001252538.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005169.mm.1 |
NM_028412.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005169.mm.1 |
NM_028412.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005169.mm.1 |
TC0200000577.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200005165.mm.1 |
NM_001252534.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005165.mm.1 |
NM_001252534.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005165.mm.1 |
NM_001252536.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005165.mm.1 |
NM_001252536.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005165.mm.1 |
NM_001252537.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005165.mm.1 |
NM_001252537.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005165.mm.1 |
NM_001252538.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005165.mm.1 |
NM_001252538.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005165.mm.1 |
NM_028412.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005165.mm.1 |
NM_028412.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005165.mm.1 |
TC0200000577.mm.1 |
MTA TC |