Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200004815.mm.1 |
NM_001112703.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004815.mm.1 |
NM_001283046.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004815.mm.1 |
NM_001283047.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004815.mm.1 |
TC0200000558.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200004820.mm.1 |
NM_001283047.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004820.mm.1 |
TC0200000558.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200004827.mm.1 |
NM_001283045.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004827.mm.1 |
TC0200000558.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200004816.mm.1 |
NM_001112703.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004816.mm.1 |
NM_001283046.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004816.mm.1 |
NM_001283047.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004816.mm.1 |
TC0200000558.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200004817.mm.1 |
NM_001112703.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004817.mm.1 |
NM_001283046.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004817.mm.1 |
NM_001283047.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004817.mm.1 |
TC0200000558.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200004813.mm.1 |
NM_001112703.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004813.mm.1 |
NM_001283046.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004813.mm.1 |
NM_001283047.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004813.mm.1 |
TC0200000558.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200004818.mm.1 |
NM_001112703.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004818.mm.1 |
NM_001283046.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004818.mm.1 |
NM_001283047.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004818.mm.1 |
TC0200000558.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200004831.mm.1 |
NM_001112703.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004831.mm.1 |
NM_001283045.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004831.mm.1 |
NM_001283046.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004831.mm.1 |
NM_001283047.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004831.mm.1 |
NM_009594.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004831.mm.1 |
TC0200000558.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200004819.mm.1 |
NM_001283046.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004819.mm.1 |
TC0200000558.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200004832.mm.1 |
NM_001112703.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004832.mm.1 |
NM_001283045.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004832.mm.1 |
NM_001283046.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004832.mm.1 |
NM_001283047.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004832.mm.1 |
NM_009594.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200004832.mm.1 |
TC0200000558.mm.1 |
MTA TC |