Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164357.1 | NP_001157829.1 | 1 - 2 | 398 | 99 | |
NM_146118.3 | NP_666230.2 | 1 - 2 | 398 | 99 | ||
GenBank mRNA | AK049392.1 | - | 1 - 2 | 374 | 99 | |
AK082756.1 | - | 1 - 2 | 355 | 99 | ||
AK132204.1 | - | 1 - 2 | 352 | 99 | ||
BB653459.1 | - | 1 - 2 | 399 | 99 | ||
BC037109.1 | - | 1 - 2 | 358 | 5368306 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |