Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164274.1 | NP_001157746.1 | 4 - 5 | 2268 | 104 | |
NM_013720.2 | NP_038748.2 | 4 - 5 | 2268 | 104 | ||
GenBank mRNA | AF205935.1 | - | 3 - 4 | 2082 | 104 | |
AK032570.1 | - | 4 - 5 | 2249 | 104 | ||
AK046252.1 | - | 4 - 5 | 2269 | 104 | ||
AK172962.1 | - | 4 - 5 | 2191 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |