Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030112.2 | NP_084388.2 | 7 - 8 | 1039 | 66 | |
GenBank mRNA | AK135633.1 | - | 1 - 2 | 107 | 66 | |
BC117848.1 | - | 7 - 8 | 891 | 40093717 | 66 | |
BC117849.1 | - | 7 - 8 | 891 | 40093724 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |