Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ltk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008523.2 | NP_032549.2 | 2 - 3 | 376 | 79 | |
NM_203345.2 | NP_976220.2 | 8 - 9 | 1222 | 79 | ||
NM_206941.1 | NP_996824.1 | 3 - 4 | 559 | 79 | ||
NM_206942.2 | NP_996825.2 | 7 - 8 | 1039 | 79 | ||
GenBank mRNA | BC047138.1 | - | 2 - 3 | 376 | 6389834 | 79 |
BC055898.1 | - | 2 - 3 | 370 | 79 | ||
BC141299.1 | - | 7 - 8 | 1015 | 9055911 | 79 | |
M90470.1 | - | 8 - 9 | 1222 | 79 | ||
X07984.1 | - | 2 - 3 | 350 | 79 | ||
X52621.1 | - | 2 - 3 | 365 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |