Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfkp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291071.1 | NP_001278000.1 | 3 - 4 | 355 | 61 | |
NM_019703.4 | NP_062677.1 | 3 - 4 | 355 | 61 | ||
GenBank mRNA | AF123533.1 | - | 3 - 4 | 320 | 61 | |
AK004790.1 | - | 3 - 4 | 310 | 61 | ||
AK020367.1 | - | 3 - 4 | 310 | 61 | ||
AK049841.1 | - | 3 - 4 | 309 | 61 | ||
AK054193.1 | - | 3 - 4 | 310 | 61 | ||
AK076803.1 | - | 3 - 4 | 310 | 61 | ||
AK078254.1 | - | 3 - 4 | 310 | 61 | ||
AK098162.1 | - | 3 - 4 | 309 | 61 | ||
AK152670.1 | - | 3 - 4 | 310 | 61 | ||
AK154125.1 | - | 3 - 4 | 310 | 61 | ||
AK165422.1 | - | 3 - 4 | 309 | 61 | ||
AK165425.1 | - | 3 - 4 | 309 | 61 | ||
AK170624.1 | - | 3 - 4 | 310 | 61 | ||
AK171062.1 | - | 3 - 4 | 310 | 61 | ||
AY779275.1 | - | 3 - 4 | 272 | 61 | ||
AY779276.1 | - | 3 - 4 | 272 | 61 | ||
BC006926.1 | - | 3 - 4 | 294 | 2655383 | 61 | |
BY243493.1 | - | 3 - 4 | 354 | 61 | ||
CT010268.1 | - | 3 - 4 | 263 | 61 | ||
Y19008.1 | - | 3 - 4 | 312 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |