Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Meis2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136072.2 | NP_001129544.1 | 4 - 5 | 685 | 130 | |
NM_001159567.1 | NP_001153039.1 | 4 - 5 | 685 | 130 | ||
NM_001159568.1 | NP_001153040.1 | 4 - 5 | 685 | 130 | ||
NM_001159569.1 | NP_001153041.1 | 4 - 5 | 958 | 130 | ||
NM_001159570.1 | NP_001153042.1 | 4 - 5 | 958 | 130 | ||
NM_010825.3 | NP_034955.1 | 4 - 5 | 685 | 130 | ||
GenBank mRNA | AJ000504.1 | - | 4 - 5 | 674 | 130 | |
AJ000505.1 | - | 4 - 5 | 674 | 130 | ||
AJ000506.1 | - | 4 - 5 | 674 | 130 | ||
AJ000507.1 | - | 4 - 5 | 674 | 130 | ||
AK040611.1 | - | 2 - 3 | 966 | 130 | ||
AK045126.1 | - | 2 - 3 | 430 | 130 | ||
AK146633.1 | - | 4 - 5 | 685 | 130 | ||
AK158513.1 | - | 4 - 5 | 850 | 130 | ||
AK158839.1 | - | 4 - 5 | 957 | 130 | ||
BC017375.1 | - | 4 - 5 | 751 | 4191098 | 130 | |
KM974644.1 | - | 2 - 3 | 168 | 130 | ||
U57343.1 | - | 722 | 130 | |||
U68383.1 | - | 4 - 5 | 644 | 130 | ||
U68384.1 | - | 4 - 5 | 644 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |