Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Meis2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136072.2 | NP_001129544.1 | 5 - 6 | 733 | 110 | |
NM_001159567.1 | NP_001153039.1 | 5 - 6 | 733 | 110 | ||
NM_001159568.1 | NP_001153040.1 | 5 - 6 | 733 | 110 | ||
NM_001159569.1 | NP_001153041.1 | 5 - 6 | 1006 | 110 | ||
NM_001159570.1 | NP_001153042.1 | 5 - 6 | 1006 | 110 | ||
NM_010825.3 | NP_034955.1 | 5 - 6 | 733 | 110 | ||
GenBank mRNA | AJ000504.1 | - | 5 - 6 | 722 | 110 | |
AJ000505.1 | - | 5 - 6 | 722 | 110 | ||
AJ000506.1 | - | 5 - 6 | 722 | 110 | ||
AJ000507.1 | - | 5 - 6 | 722 | 110 | ||
AK040611.1 | - | 3 - 4 | 1014 | 110 | ||
AK045126.1 | - | 3 - 4 | 478 | 110 | ||
AK146633.1 | - | 5 - 6 | 733 | 110 | ||
AK158513.1 | - | 5 - 6 | 898 | 110 | ||
AK158839.1 | - | 5 - 6 | 1005 | 110 | ||
BC017375.1 | - | 5 - 6 | 799 | 4191098 | 110 | |
U57343.1 | - | 770 | 110 | |||
U68383.1 | - | 5 - 6 | 692 | 110 | ||
U68384.1 | - | 5 - 6 | 692 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |