Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il13ra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK089687.2 | - | 6 - 7 | 1064 | 158 | |
BC003723.1 | - | 3 - 4 | 240 | 3486223 | 158 | |
U65747.1 | - | 3 - 4 | 328 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |