Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tc2n | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082976.2 | NP_001076445.1 | 10 - 11 | 1318 | 70 | |
NM_001286364.1 | NP_001273293.1 | 10 - 11 | 1981 | 70 | ||
NM_028924.3 | NP_083200.1 | 10 - 11 | 1670 | 70 | ||
GenBank mRNA | AB062282.1 | - | 9 - 10 | 1150 | 70 | |
AK016683.1 | - | 10 - 11 | 2002 | 70 | ||
AK136558.1 | - | 10 - 11 | 1319 | 70 | ||
BC120793.1 | - | 10 - 11 | 1670 | 40129716 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |