Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023668.2 | NP_076157.2 | 5 - 6 | 724 | 75 | |
GenBank mRNA | AF290472.1 | - | 5 - 6 | 656 | 75 | |
AF323918.1 | - | 5 - 6 | 744 | 75 | ||
AK011168.1 | - | 5 - 6 | 634 | 75 | ||
AK146029.1 | - | 5 - 6 | 653 | 75 | ||
BC021434.1 | - | 620 | 5029832 | 75 | ||
BC046796.1 | - | 5 - 6 | 655 | 5718032 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |