Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177779.3 | NP_808447.2 | 4 - 5 | 672 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |