Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300002566.mm.1 |
NM_001039188.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002566.mm.1 |
NM_001039188.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002566.mm.1 |
NM_001177868.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002566.mm.1 |
NM_001177868.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002566.mm.1 |
NM_001177869.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002566.mm.1 |
NM_001177869.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002566.mm.1 |
NM_026830.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002566.mm.1 |
NM_026830.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002566.mm.1 |
NR_033218.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002566.mm.1 |
NR_033218.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002566.mm.1 |
NR_033615.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002566.mm.1 |
NR_033615.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002566.mm.1 |
TC1300000432.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300002554.mm.1 |
NM_001177868.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002554.mm.1 |
NM_001177868.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002554.mm.1 |
NR_033615.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002554.mm.1 |
NR_033615.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002554.mm.1 |
TC1300000432.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300002564.mm.1 |
NM_001039188.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002564.mm.1 |
NM_001039188.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002564.mm.1 |
NM_001177868.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002564.mm.1 |
NM_001177868.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002564.mm.1 |
NM_001177869.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002564.mm.1 |
NM_001177869.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002564.mm.1 |
NM_026830.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002564.mm.1 |
NM_026830.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002564.mm.1 |
NR_033218.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002564.mm.1 |
NR_033218.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002564.mm.1 |
NR_033615.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002564.mm.1 |
NR_033615.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002564.mm.1 |
TC1300000432.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300002556.mm.1 |
NM_001039188.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002556.mm.1 |
NM_001039188.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002556.mm.1 |
TC1300000432.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300002561.mm.1 |
NM_001039188.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002561.mm.1 |
NM_001039188.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002561.mm.1 |
NM_001177868.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002561.mm.1 |
NM_001177868.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002561.mm.1 |
NM_001177869.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002561.mm.1 |
NM_001177869.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002561.mm.1 |
NM_026830.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002561.mm.1 |
NM_026830.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002561.mm.1 |
NR_033218.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002561.mm.1 |
NR_033218.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002561.mm.1 |
NR_033615.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002561.mm.1 |
NR_033615.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002561.mm.1 |
TC1300000432.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300002563.mm.1 |
NM_001039188.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002563.mm.1 |
NM_001039188.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002563.mm.1 |
NM_001177868.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002563.mm.1 |
NM_001177868.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002563.mm.1 |
NM_001177869.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002563.mm.1 |
NM_001177869.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002563.mm.1 |
NM_026830.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002563.mm.1 |
NM_026830.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002563.mm.1 |
NR_033218.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002563.mm.1 |
NR_033218.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002563.mm.1 |
NR_033615.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002563.mm.1 |
NR_033615.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002563.mm.1 |
TC1300000432.mm.1 |
MTA TC |