Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wwc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170779.1 | NP_740749.1 | 11 - 12 | 1813 | 81 | |
GenBank mRNA | AB271907.1 | - | 11 - 12 | 1783 | 81 | |
AK220259.1 | - | 2184 | 81 | |||
BC006733.1 | - | 8 - 9 | 1402 | 81 | ||
BC017638.1 | - | 4 - 5 | 939 | 81 | ||
BC037006.1 | - | 3 - 4 | 661 | 5116674 | 81 | |
DQ256090.1 | - | 11 - 12 | 1813 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |