Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp6nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080548.1 | NP_001074017.1 | 1 - 2 | 194 | 150 | |
GenBank mRNA | AK155073.1 | - | 1 - 2 | 195 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease Mus musculus Probe spans exons |