Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200015912.mm.1 |
NM_001166350.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015912.mm.1 |
NM_001166350.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015912.mm.1 |
NM_199314.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015912.mm.1 |
NM_199314.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015912.mm.1 |
TC1200002535.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200015923.mm.1 |
NM_199314.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015923.mm.1 |
NM_199314.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015923.mm.1 |
TC1200002535.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200015914.mm.1 |
NM_001166350.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015914.mm.1 |
NM_001166350.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015914.mm.1 |
NM_199314.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015914.mm.1 |
NM_199314.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015914.mm.1 |
TC1200002535.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200015913.mm.1 |
NM_001166350.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015913.mm.1 |
NM_001166350.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015913.mm.1 |
NM_199314.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015913.mm.1 |
NM_199314.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015913.mm.1 |
TC1200002535.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200015917.mm.1 |
NM_001166350.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015917.mm.1 |
NM_001166350.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015917.mm.1 |
NM_199314.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015917.mm.1 |
NM_199314.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015917.mm.1 |
TC1200002535.mm.1 |
MTA TC |