Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Leng8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172736.3 | NP_766324.1 | 1 - 2 | 89 | 63 | |
GenBank mRNA | AK034310.1 | - | 1 - 2 | 94 | 63 | |
AK085304.1 | - | 1 - 2 | 87 | 63 | ||
AK170629.1 | - | 1 - 2 | 97 | 63 | ||
BC041775.1 | - | 1 - 2 | 61 | 63 | ||
BC042658.1 | - | 1 - 2 | 77 | 5365041 | 63 | |
BC066768.1 | - | 1 - 2 | 80 | 6407146 | 63 | |
CF536188.1 | - | 1 - 2 | 89 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |