Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100010550.mm.1 |
NM_033475.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010550.mm.1 |
NM_033475.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010550.mm.1 |
TC1100001149.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100010569.mm.1 |
NM_001159482.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010569.mm.1 |
NM_001159482.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010569.mm.1 |
NM_033475.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010569.mm.1 |
NM_033475.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010569.mm.1 |
TC1100001149.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100010549.mm.1 |
NM_033475.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010549.mm.1 |
NM_033475.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010549.mm.1 |
TC1100001149.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100010552.mm.1 |
NM_001159482.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010552.mm.1 |
NM_001159482.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010552.mm.1 |
NM_033475.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010552.mm.1 |
NM_033475.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010552.mm.1 |
TC1100001149.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100010561.mm.1 |
NM_001159482.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010561.mm.1 |
NM_001159482.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010561.mm.1 |
NM_033475.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010561.mm.1 |
NM_033475.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010561.mm.1 |
TC1100001149.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100010562.mm.1 |
NM_001159482.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010562.mm.1 |
NM_001159482.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010562.mm.1 |
NM_033475.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010562.mm.1 |
NM_033475.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010562.mm.1 |
TC1100001149.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100010551.mm.1 |
NM_033475.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010551.mm.1 |
NM_033475.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010551.mm.1 |
TC1100001149.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100010553.mm.1 |
NM_001159482.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010553.mm.1 |
NM_001159482.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010553.mm.1 |
NM_033475.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010553.mm.1 |
NM_033475.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010553.mm.1 |
TC1100001149.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100010554.mm.1 |
NM_001159482.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010554.mm.1 |
NM_001159482.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010554.mm.1 |
NM_033475.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010554.mm.1 |
NM_033475.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010554.mm.1 |
TC1100001149.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100010556.mm.1 |
NM_001159482.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010556.mm.1 |
NM_001159482.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010556.mm.1 |
NM_033475.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010556.mm.1 |
NM_033475.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010556.mm.1 |
TC1100001149.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100010548.mm.1 |
NM_001159482.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010548.mm.1 |
NM_001159482.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010548.mm.1 |
TC1100001149.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100010557.mm.1 |
NM_001159482.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010557.mm.1 |
NM_001159482.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010557.mm.1 |
NM_033475.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010557.mm.1 |
NM_033475.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010557.mm.1 |
TC1100001149.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100010567.mm.1 |
NM_001159482.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010567.mm.1 |
NM_001159482.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010567.mm.1 |
NM_033475.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010567.mm.1 |
NM_033475.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010567.mm.1 |
TC1100001149.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100010560.mm.1 |
NM_001159482.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010560.mm.1 |
NM_001159482.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010560.mm.1 |
NM_033475.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010560.mm.1 |
NM_033475.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010560.mm.1 |
TC1100001149.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100010564.mm.1 |
NM_001159482.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010564.mm.1 |
NM_001159482.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010564.mm.1 |
NM_033475.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010564.mm.1 |
NM_033475.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100010564.mm.1 |
TC1100001149.mm.1 |
MTA TC |