Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163645.1 | NP_001157117.1 | 15 - 16 | 2265 | 68 | |
NM_027881.3 | NP_082157.2 | 15 - 16 | 1983 | 68 | ||
GenBank mRNA | AK004768.1 | - | 15 - 16 | 1985 | 68 | |
AK037114.1 | - | 15 - 16 | 1952 | 68 | ||
BC044866.1 | - | 14 - 15 | 1910 | 5003631 | 68 | |
BC086485.1 | - | 15 - 16 | 2177 | 5344992 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |