Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600004832.mm.1 |
TC0600000633.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600004832.mm.1 |
NM_025816.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004832.mm.1 |
NM_025816.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004841.mm.1 |
TC0600000633.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600004841.mm.1 |
NM_025816.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004841.mm.1 |
NM_025816.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004839.mm.1 |
TC0600000633.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600004839.mm.1 |
NM_025816.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004839.mm.1 |
NM_025816.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004833.mm.1 |
TC0600000633.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600004833.mm.1 |
NM_025816.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004833.mm.1 |
NM_025816.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004838.mm.1 |
TC0600000633.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600004838.mm.1 |
NM_025816.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004838.mm.1 |
NM_025816.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004844.mm.1 |
TC0600000633.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600004844.mm.1 |
NM_025816.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004844.mm.1 |
NM_025816.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004845.mm.1 |
TC0600000633.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600004845.mm.1 |
NM_025816.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004845.mm.1 |
NM_025816.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004843.mm.1 |
TC0600000633.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600004843.mm.1 |
NM_025816.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004843.mm.1 |
NM_025816.3 |
Refseq |