Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scaf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134123.3 | NP_598884.2 | 6 - 7 | 1021 | 77 | |
GenBank mRNA | AK122451.1 | - | 6 - 7 | 888 | 77 | |
AK157957.1 | - | 6 - 7 | 1027 | 77 | ||
AK171078.1 | - | 7 - 8 | 828 | 77 | ||
AK218201.1 | - | 2 - 3 | 77 | |||
BC038363.1 | - | 6 - 7 | 665 | 5371946 | 77 | |
BC075621.1 | - | 6 - 7 | 1014 | 6406441 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |