Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sel1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039089.1 | NP_001034178.1 | 16 - 17 | 1740 | 59 | |
NM_011344.2 | NP_035474.1 | 16 - 17 | 1590 | 59 | ||
GenBank mRNA | AF063095.1 | - | 16 - 17 | 1539 | 59 | |
AK005023.2 | - | 16 - 17 | 1713 | 59 | ||
AK146022.1 | - | 16 - 17 | 1714 | 59 | ||
AK160936.1 | - | 16 - 17 | 1548 | 59 | ||
AK220502.1 | - | 16 - 17 | 1740 | 59 | ||
BC026816.1 | - | 16 - 17 | 1562 | 4457637 | 59 | |
BC049959.1 | - | 11 - 12 | 952 | 59 | ||
BC057452.1 | - | 2 - 3 | 175 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |