Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sel1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039089.1 | NP_001034178.1 | 8 - 9 | 1010 | 83 | |
NM_011344.2 | NP_035474.1 | 8 - 9 | 860 | 83 | ||
GenBank mRNA | AF063095.1 | - | 8 - 9 | 809 | 83 | |
AK005023.2 | - | 8 - 9 | 983 | 83 | ||
AK146022.1 | - | 8 - 9 | 984 | 83 | ||
AK160936.1 | - | 8 - 9 | 818 | 83 | ||
AK161546.1 | - | 8 - 9 | 984 | 83 | ||
AK220502.1 | - | 8 - 9 | 1010 | 83 | ||
BC026816.1 | - | 8 - 9 | 832 | 4457637 | 83 | |
BC049959.1 | - | 3 - 4 | 222 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |