Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600004994.mm.1 |
TC0600000651.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600004994.mm.1 |
NM_001167860.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004994.mm.1 |
NM_001167860.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004994.mm.1 |
NM_001167861.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004994.mm.1 |
NM_001167861.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004995.mm.1 |
TC0600000651.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600004995.mm.1 |
NM_001167860.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004995.mm.1 |
NM_001167860.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004995.mm.1 |
NM_001167861.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004995.mm.1 |
NM_001167861.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004992.mm.1 |
TC0600000651.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600004992.mm.1 |
NM_001167860.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004992.mm.1 |
NM_001167860.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004996.mm.1 |
TC0600000651.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600004996.mm.1 |
NM_001167860.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004996.mm.1 |
NM_001167860.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004996.mm.1 |
NM_001167861.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600004996.mm.1 |
NM_001167861.1 |
Refseq |