Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163640.1 | NP_001157112.1 | 6 - 7 | 747 | 66 | |
GenBank mRNA | AK131641.1 | - | 6 - 7 | 641 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |