Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163640.1 | NP_001157112.1 | 9 - 10 | 1088 | 118 | |
NM_023543.2 | NP_076032.2 | 3 - 4 | 591 | 118 | ||
GenBank mRNA | AJ279014.1 | - | 4 - 5 | 489 | 118 | |
AK006398.1 | - | 4 - 5 | 685 | 118 | ||
AK131641.1 | - | 9 - 10 | 982 | 118 | ||
AK160255.1 | - | 4 - 5 | 420 | 118 | ||
BC051139.1 | - | 3 - 4 | 592 | 6589539 | 118 | |
BC141060.1 | - | 3 - 4 | 550 | 9055672 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |