Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028846.5 | NP_083122.1 | 1 - 2 | 117 | 73 | |
GenBank mRNA | AF449715.1 | - | 1 - 2 | 68 | 73 | |
AK054279.1 | - | 1 - 2 | 60 | 73 | ||
BY251374.1 | - | 1 - 2 | 118 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mus musculus Probe spans exons |