Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301781.1 | NP_001288710.1 | 1 - 2 | 295 | 128 | |
NM_207232.2 | NP_997115.1 | 1 - 2 | 295 | 128 | ||
GenBank mRNA | AK030415.1 | - | 1 - 2 | 288 | 128 | |
AK053480.1 | - | 1 - 2 | 133 | 128 | ||
AK132781.1 | - | 1 - 2 | 204 | 128 | ||
BC066081.1 | - | 1 - 2 | 196 | 30355705 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |