Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008498.2 | NP_001008498.2 | 6 - 7 | 990 | 109 | |
GenBank mRNA | AK014304.1 | - | 6 - 7 | 989 | 109 | |
BC085113.1 | - | 6 - 7 | 946 | 30730999 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |