Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200006757.mm.1 |
NM_175238.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006757.mm.1 |
TC0200000821.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006761.mm.1 |
NM_175238.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006761.mm.1 |
TC0200000821.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006755.mm.1 |
NM_175238.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006755.mm.1 |
TC0200000821.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006754.mm.1 |
NM_175238.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006754.mm.1 |
TC0200000821.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006753.mm.1 |
NM_175238.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006753.mm.1 |
TC0200000821.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006758.mm.1 |
NM_175238.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006758.mm.1 |
TC0200000821.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006760.mm.1 |
NM_175238.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006760.mm.1 |
TC0200000821.mm.1 |
MTA TC |