Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175238.5 | NP_780447.4 | 28 - 29 | 3367 | 97 | |
GenBank mRNA | AK017618.1 | - | 2 - 3 | 125 | 97 | |
AY428571.2 | - | 28 - 29 | 3351 | 97 | ||
AY585206.1 | - | 27 - 28 | 3360 | 97 | ||
JX255737.1 | - | 27 - 28 | 3303 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |