Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pacs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081170.1 | NP_001074639.1 | 15 - 16 | 1678 | 77 | |
NM_001291444.1 | NP_001278373.1 | 15 - 16 | 1684 | 77 | ||
NM_001291445.1 | NP_001278374.1 | 14 - 15 | 1588 | 77 | ||
GenBank mRNA | AK030003.1 | - | 10 - 11 | 1047 | 77 | |
AK036581.1 | - | 14 - 15 | 1564 | 77 | ||
AK054000.1 | - | 15 - 16 | 1659 | 77 | ||
AK122326.1 | - | 13 - 14 | 1367 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Probe spans exons |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2218223 | BC043302.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2218223 | BC059076.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2218223 | AK054000.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2218223 | AK122326.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2218223 | AK030003.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 860659 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 192451 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 885557 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 947900 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 96486 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 823729 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 799720 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 109413 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 500867 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914407 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 790421 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 962423 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 630496 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 21617 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904995 | AK013106.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904995 | AK044792.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904995 | AK076018.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904995 | AK088887.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904995 | AK132779.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904995 | AK135173.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904995 | AK166045.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904995 | AK166698.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904995 | BC046554.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904995 | BC062916.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904995 | BC086320.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904995 | BC108369.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904995 | BC129873.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904995 | BC129874.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904995 | NM_007840.3 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904995 | X65627.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 543685 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510118 | AK047380.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510118 | AK155135.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510118 | U42384.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 176682 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 921850 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 422716 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 842268 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 941171 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 152620 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 826506 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 138630 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 246320 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 426244 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 221884 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 382969 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 863403 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 150860 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 920432 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 246472 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 522967 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 35007 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 682340 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 564071 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 143809 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 592823 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 444021 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 93827 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 46889 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 38041 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 89169 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 87612 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143783 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P108761 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105282 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2218223 | BC043302.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2218223 | BC059076.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2218223 | AK054000.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2218223 | AK122326.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2218223 | AK030003.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P390446 | BC043302.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P390446 | BC051499.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P390446 | BC059076.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P390446 | AK122326.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P390446 | AK216671.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P439520 | BC043302.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P439520 | BC059076.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P439520 | AK054000.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P439520 | AK122326.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P439520 | AK030003.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P213004 | AK054000.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P213004 | AK122326.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P213004 | AK030003.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P213004 | AK036581.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P213004 | NM_001081170.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126089 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121346 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09985026 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09985008 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09985028 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09985003 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09985020 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10398955 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10398948 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10398942 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10398943 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10398961 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10398939 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10398953 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10398944 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10398950 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10398946 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10398962 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10398958 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10398957 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10398945 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10398951 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10398959 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10398947 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10398937 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10398941 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10398960 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10398963 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10398956 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10398952 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10398949 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10398954 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10398964 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10398938 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10398965 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10398940 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1426963_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1434255_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10398936 | - | - |
Mouse Genome 430 2.0 Array | 1426963_at | - | - |
Mouse Genome 430 2.0 Array | 1434255_at | - | - |
Mouse Genome 430 2.0 Array | 1426963_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1434255_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 217893_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 217893_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1200009210.mm.1 | NM_001081170.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200009210.mm.1 | NM_001081170.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200009210.mm.1 | TC1200001277.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200009216.mm.1 | NM_001081170.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200009216.mm.1 | NM_001081170.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200009216.mm.1 | TC1200001277.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200009218.mm.1 | NM_001081170.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200009218.mm.1 | NM_001081170.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200009218.mm.1 | TC1200001277.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200009213.mm.1 | NM_001081170.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200009213.mm.1 | NM_001081170.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200009213.mm.1 | TC1200001277.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200009209.mm.1 | NM_001081170.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200009209.mm.1 | NM_001081170.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200009209.mm.1 | TC1200001277.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200009211.mm.1 | NM_001081170.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200009211.mm.1 | NM_001081170.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200009211.mm.1 | TC1200001277.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200009215.mm.1 | NM_001081170.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200009215.mm.1 | NM_001081170.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200009215.mm.1 | TC1200001277.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200009217.mm.1 | NM_001081170.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200009217.mm.1 | NM_001081170.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200009217.mm.1 | TC1200001277.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200009219.mm.1 | NM_001081170.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200009219.mm.1 | NM_001081170.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200009219.mm.1 | TC1200001277.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200009212.mm.1 | NM_001081170.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200009212.mm.1 | NM_001081170.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200009212.mm.1 | TC1200001277.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200009214.mm.1 | NM_001081170.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200009214.mm.1 | NM_001081170.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200009214.mm.1 | TC1200001277.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200009220.mm.1 | NM_001081170.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200009220.mm.1 | NM_001081170.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200009220.mm.1 | TC1200001277.mm.1 | MTA TC |
Mouse Expression Set 430 | 1426963_PM_at | - | - |
Mouse Expression Set 430 | 1426963_at | - | - |
Mouse Expression Set 430 | 1434255_PM_at | - | - |
Mouse Expression Set 430 | 1434255_at | - | - |
Murine Genome U74v2 Set | 171358_PM_i_at | - | - |
Murine Genome U74v2 Set | 171358_i_at | - | - |
Mouse Genome 430A 2.0 Array | 1426963_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1426963_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1426963_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1426963_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1434255_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1434255_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 103140301 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2620583 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2620583 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1259927 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 103140301 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103140301 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1259927 | - | - |