Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054081.2 | NP_473422.2 | 12 - 13 | 1283 | 133 | |
GenBank mRNA | AB794093.1 | - | 13 - 14 | 1194 | 133 | |
AB794094.1 | - | 12 - 13 | 1143 | 133 | ||
AB794096.1 | - | 13 - 14 | 1194 | 133 | ||
AB794098.1 | - | 12 - 13 | 1143 | 133 | ||
AB794099.1 | - | 12 - 13 | 1143 | 133 | ||
AB794101.1 | - | 13 - 14 | 1194 | 133 | ||
AB794103.1 | - | 12 - 13 | 1143 | 133 | ||
AF288137.1 | - | 12 - 13 | 1245 | 133 | ||
AF463504.1 | - | 13 - 14 | 1194 | 133 | ||
BC024527.1 | - | 5 - 6 | 509 | 133 | ||
BC112910.1 | - | 13 - 14 | 1334 | 6515946 | 133 | |
CJ139622.1 | - | 4 - 5 | 434 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |