Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081033.1 | NP_001074502.1 | 2 - 3 | 1112 | 109 | |
GenBank mRNA | BC158125.1 | - | 2 - 3 | 1252 | 9088197 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |