Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dolpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290508.1 | NP_001277437.1 | 6 - 7 | 846 | 60 | |
NM_020329.4 | NP_065062.1 | 5 - 6 | 639 | 60 | ||
GenBank mRNA | AB030189.1 | - | 5 - 6 | 479 | 60 | |
AK002544.1 | - | 6 - 7 | 685 | 60 | ||
AK010797.1 | - | 1 - 2 | 76 | 60 | ||
AK044141.1 | - | 5 - 6 | 478 | 60 | ||
AK053923.1 | - | 5 - 6 | 468 | 60 | ||
AK088065.1 | - | 5 - 6 | 483 | 60 | ||
BC026544.1 | - | 5 - 6 | 463 | 4975127 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |