Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010230.2 | NP_034360.2 | 2 - 3 | 2089 | 109 | |
GenBank mRNA | X53599.1 | - | 2 - 3 | 2043 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |