Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146252.2 | NP_666364.1 | 7 - 8 | 563 | 67 | |
GenBank mRNA | AK158066.1 | - | 7 - 8 | 644 | 67 | |
BC025586.1 | - | 7 - 8 | 589 | 5324875 | 67 | |
BC039215.1 | - | 1 - 2 | 152 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |