Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029148.1 | NP_083424.1 | 2 - 3 | 393 | 68 | |
GenBank mRNA | AK015667.1 | - | 2 - 3 | 405 | 68 | |
AK030696.1 | - | 2 - 3 | 407 | 68 | ||
AK033111.1 | - | 2 - 3 | 315 | 68 | ||
AK040779.1 | - | 2 - 3 | 378 | 68 | ||
AK053897.1 | - | 2 - 3 | 2524 | 68 | ||
AK129301.1 | - | 2 - 3 | 392 | 68 | ||
AK147465.1 | - | 2 - 3 | 394 | 68 | ||
BC050918.1 | - | 2 - 3 | 292 | 68 | ||
BC054558.1 | - | 2 - 3 | 393 | 6405599 | 68 | |
BC138168.1 | - | 2 - 3 | 367 | 8861187 | 68 | |
BC138169.1 | - | 2 - 3 | 367 | 8861188 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |