Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153805.1 | NP_722500.1 | 3 - 4 | 407 | 128 | |
GenBank mRNA | BC034126.1 | - | 3 - 4 | 407 | 4912536 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |