Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135657.1 | NP_001129129.1 | 3 - 4 | 477 | 105 | |
NM_008982.5 | NP_033008.3 | 3 - 4 | 761 | 105 | ||
GenBank mRNA | AK147318.1 | - | 3 - 4 | 682 | 105 | |
AK147556.1 | - | 3 - 4 | 682 | 105 | ||
AY038891.1 | - | 3 - 4 | 579 | 105 | ||
D45212.1 | - | 3 - 4 | 717 | 105 | ||
DQ133576.1 | - | 3 - 4 | 492 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |