Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009537.3 | NP_033563.2 | 1 - 2 | 773 | 82 | |
GenBank mRNA | AK076149.1 | - | 2 - 3 | 386 | 82 | |
AK166542.1 | - | 2 - 3 | 111 | 82 | ||
BC055899.1 | - | 1 - 2 | 751 | 3156776 | 82 | |
M73963.1 | - | 1 - 2 | 773 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |