Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssc5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173008.2 | NP_766596.1 | 6 - 7 | 1008 | 65 | |
GenBank mRNA | AK079906.1 | - | 6 - 7 | 1009 | 65 | |
AK165666.1 | - | 6 - 7 | 1070 | 65 | ||
BC141044.1 | - | 6 - 7 | 1006 | 9055656 | 65 | |
EU850434.1 | - | 6 - 7 | 1009 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |