Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssc5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173008.2 | NP_766596.1 | 8 - 9 | 1492 | 73 | |
GenBank mRNA | AK079906.1 | - | 8 - 9 | 1493 | 73 | |
AK165666.1 | - | 8 - 9 | 1554 | 73 | ||
BC141044.1 | - | 8 - 9 | 1490 | 9055656 | 73 | |
EU850434.1 | - | 8 - 9 | 1493 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |