Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpnmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053110.4 | NP_444340.3 | 7 - 8 | 1368 | 114 | |
GenBank mRNA | AF322054.1 | - | 7 - 8 | 1206 | 114 | |
AJ251685.1 | - | 7 - 8 | 1253 | 114 | ||
AK044764.1 | - | 7 - 8 | 1262 | 114 | ||
AK076347.1 | - | 7 - 8 | 1237 | 114 | ||
AK079220.1 | - | 1210 | 114 | |||
AK149740.1 | - | 7 - 8 | 1276 | 114 | ||
AK150739.1 | - | 7 - 8 | 1237 | 114 | ||
AK154617.1 | - | 7 - 8 | 1272 | 114 | ||
AK159165.1 | - | 7 - 8 | 1267 | 114 | ||
AK159270.1 | - | 7 - 8 | 1275 | 114 | ||
AK159747.1 | - | 7 - 8 | 1237 | 114 | ||
AK159908.1 | - | 7 - 8 | 1242 | 113 | ||
AK170506.1 | - | 7 - 8 | 1267 | 114 | ||
AK171347.1 | - | 7 - 8 | 1243 | 114 | ||
AK171441.1 | - | 7 - 8 | 1237 | 114 | ||
AK171465.1 | - | 7 - 8 | 1237 | 114 | ||
AK171474.1 | - | 7 - 8 | 1237 | 114 | ||
AK171517.1 | - | 7 - 8 | 1355 | 114 | ||
AK171608.1 | - | 7 - 8 | 1237 | 114 | ||
AK171618.1 | - | 7 - 8 | 1237 | 114 | ||
BC026375.1 | - | 7 - 8 | 1242 | 4164706 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |