Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291049.1 | NP_001277978.1 | 2 - 3 | 738 | 105 | |
NM_173754.4 | NP_776115.2 | 2 - 3 | 738 | 105 | ||
GenBank mRNA | AK042217.1 | - | 2 - 3 | 739 | 105 | |
AK083271.1 | - | 2 - 3 | 576 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mus musculus Inventoried Probe spans exons |