Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163701.1 | NP_001157173.1 | 21 - 22 | 3265 | 88 | |
NM_177294.5 | NP_796268.3 | 22 - 23 | 3342 | 88 | ||
GenBank mRNA | AK122505.1 | - | 22 - 23 | 3309 | 88 | |
AK154007.1 | - | 21 - 22 | 3249 | 88 | ||
AK161487.1 | - | 22 - 23 | 3325 | 88 | ||
AK164658.1 | - | 22 - 23 | 3326 | 88 | ||
AK166232.1 | - | 21 - 22 | 3248 | 88 | ||
AK167589.1 | - | 21 - 22 | 3248 | 88 | ||
AK171233.1 | - | 20 - 21 | 3025 | 88 | ||
BC051680.1 | - | 8 - 9 | 1258 | 88 | ||
BC088899.1 | - | 20 - 21 | 3070 | 88 | ||
BC117973.1 | - | 21 - 22 | 3234 | 40098643 | 88 | |
BC117974.1 | - | 21 - 22 | 3234 | 40098644 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |